6-Azauracil 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

15397-0050
TS15397-0050EA 90.32 USD
TS15397-0050
6-Azauracil 99%
6-Azauracil
Formula: C₃H₃N₃O₂
MW: 113.08
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00006456
CAS Number: 461-89-2
EINECS: 207-318-0
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 276 to 280 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR