Spex NanoSNAP™ Microvolume Spectrophotometer, SPEX SamplePrep
Supplier: SPEX SamplePrep
83057-02
76547-964EA 9835.55 USD
76547-964
Spex NanoSNAP™ Microvolume Spectrophotometer, SPEX SamplePrep
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Measure small sample volumes as low as 1 μl with a high degree of accuracy, reproducibility, and speed.
  • Touch-screen interface features preprogrammed DNA, RNA, and proteins measurements protocols, with customized methods available
  • Ideal for nucleic acids and proteins measurement
  • Only 1 μl sample volume required
  • Fast measurement in 3 sec
  • Stainless steel and quartz window with hydrophobic treatment
The Spex NanoSNAP™ microvolume spectrophotometer is a fast, microvolume spectrophotometer that provides an accurate and intuitive operational experience in only 3 sec. This spectrophotometer is ideal for Nucleic Acid and Protein measurement with a 190 to 1,000 nm wavelength range. Designed with your convenience in mind, this unit features preprogrammed DNA, RNA, and proteins measurements protocols, with customized methods available.
Navigation is fast and intuitive on the large 7" high resolution color touchscreen display. The home screen provides easy access to measurement modes as well as status of the system. Easily export data via the USB Type-A port for a USB flash drive of the front of the unit. A nano hydrophobic coating layer on the quartz window protects optical fibers and reduces contamination.
Delivery information: Spectrophotometer, power adapter and cords (US, UK, EU, Australia/China), and calibration report.