Masterflex® Ismatec® Tubing Bed Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
ISM011A ISM010A
MFLX07611-10CS 696 USD
MFLX07611-10 MFLX07611-15
Masterflex® Ismatec® Tubing Bed Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Ideal for delicate substances which require gentle occlusion setting.
  • 6 rollers│Snap your pump head onto your drives quickly with turn-and-lock mounting system
  • Select from interchangeable pump heads that offer flow rates from 0.01 to 3660 ml/min
Choose from two-stop, three-stop, or nominal tubing pump heads. Spring-loaded tubing bed adjusts occlusion for optimum pressure and tubing tension. Pump head plate 78002-80 is required for pump heads 07611-05, -10, and -15.