Masterflex® Transfer Tubing, Coiled FEP, 24" Relaxed, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
43607 59325
MFLX95821-05EA 472.25 USD
MFLX95821-05 MFLX95821-03
Masterflex® Transfer Tubing, Coiled FEP, 24" Relaxed, Avantor®
Tubing Transfer Tubing
Retracts for easier handling.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Our retractable coil FEP tubing offers the chemical inertness of our standard FEP tubing combined with the flexibility of a coil. FEP tubing has excellent heat resistance, a low coefficient of friction, and won't deteriorate with age. Tubing withstands temperatures from -454 to 400 °F (-270 to 205 °C); and can be sterilized by autoclave, with ethylene oxide, or by using dry heat. Choose the size and length that best fits your application. Use tubing with compression fittings.