Masterflex® High-Flow Dual Piston Pumps, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
HR300SRX
MFLX74931-50EA 21780 USD
MFLX74931-50
Masterflex® High-Flow Dual Piston Pumps, Avantor®
Pumps Piston Pumps
High throughput workhorse performs in rough conditions.
  • Flow rate up to 300 ml/min
  • Rugged stainless steel housing protects pump in harsh environments
  • PLC compatible for industrial processes and PC compatible for laboratory applications
  • Self-flushing pump head protects piston and seals
Constant flow pump is ideal for preparative chromatography. Automatic pressure compensation and flow calibration ensure consistent and repeatable results.
Self-flushing pump head washes piston with each stroke, protecting seals from degradation and abras
Delivery information: Includes detachable power cord.