Masterflex® Adapter Fittings, Sanitary Clamp, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
3080076 3080077 3080075 3080078 3080074
MFLX30800-78EA 24.75 USD
MFLX30800-78 MFLX30800-74 MFLX30800-75 MFLX30800-76 MFLX30800-77
Masterflex® Adapter Fittings, Sanitary Clamp, Avantor®
Fittings and Connectors
Meets FDA requirements and 3A sanitary standards.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Easily connects pipes or fittings of different types
  • Ideal for pharmaceutical industry
  • Chemical compatibility for use with corrosive acids and bases
These 304 stainless steel sanitary clamps are ideal for use with the stainless steel sanitary adapters. These clamps offer durable support, while connecting two sanitary adapters, for versatile application requirements. The 304 stainless steel couplings are ideal for the food, beverage, and dairy industries. They offer good chemical compatibility and are cost effective.
Adapters and gaskets required for a complete system are sold separately.