Masterflex® Direct-Reading Variable-Area Flowmeters with Valve, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
ACRXB0V02 ACRXB0V04 ACRXB0V01 ACRXB0V05
MFLX34500-81EA 498 USD
MFLX34500-81 MFLX34500-80 MFLX34500-84 MFLX34500-83
Masterflex® Direct-Reading Variable-Area Flowmeters with Valve, Avantor®
Flow Meters
Durable valve allows for easy control of your media.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Yellow colored backing enhances readability and minimizes eye fatigue
  • Use with various media for laboratory and industrial applications
These variable-area flowmeters are offered with direct reading scales for water, argon, oxygen, carbon dioxide, nitrogen or helium. Multi-turn valve is included, and dual scales display flow rates in both metric and English units. Optional scales can be developed for diverse flow conditions facilitating OEM applications. Flowmeters are configured for in-line 3/4" NPT(F) connections.
Scales can be purchased and interchanged. To change the scales, simply remove two small screws on front plate of meter and remove the plastic cover, and replace scale with desired scale, and replace cover and screws. For more details, contact an Applications Specialist.