4-Acetamidobenzenesulfonyl azide 97%

Supplier: Thermo Scientific Chemicals
Warning

44380-0050 44380-0250
TS44380-0250EA 91.07 USD
TS44380-0250 TS44380-0050
4-Acetamidobenzenesulfonyl azide 97%
4-Acetamidobenzenesulphonyl azide

Order Now

Specification Test Results

Appearance White to grey to brown Crystalline powder and or chunks
Infrared spectrum Conforms
Melting point 107 to 111 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR