Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate, anhydrous, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
, 1-Propylphosphonic anhydride
42931-1000
TS42931-1000EA
301
USD
TS42931-1000
Propanephosphonic acid cyclic anhydride 50% (w/w) in ethyl acetate, anhydrous, AcroSeal™
Propanephosphonic acid cyclic anhydride
Formula:
C₉H₂₁O₆P₃ MW: 318.19 Boiling Pt: 77.2 °C Density: 1.069 g/cm³ Flash Pt: –5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006583 CAS Number: 68957-94-8 UN: 2924 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to amber Solution |
Assay | ≥50 % w/w Solids |
Alkalinity | ≤0.03 % (as Ca(OH)2) |
Phosphate (PO4) | ≤0.003 % |
Sulfate (SO4) | ≤0.05 % |
Iron (Fe) | ≤0.005 % |
Acidity (HCl) | ≤0.025 % |
Insoluble matter | ≤0.025 % (in water and ppt by NH4OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...