Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF, anhydrous, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4,6-Tripropyl-1,3,5,2,4,6-trioxotriphosphorinane-2,4,6-trioxide
, 1-Propylphosphonic anhydride
42911-1000
TS42911-1000EA
351
USD
TS42911-1000
Propanephosphonic acid cyclic anhydride 50% (w/w) in DMF, anhydrous, AcroSeal™
Propanephosphonic acid cyclic anhydride
Formula:
C₉H₂₁O₆P₃ MW: 318.19 Boiling Pt: 153 °C Density: 1.09 g/cm³ Flash Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006583 CAS Number: 68957-94-8 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow to brown Solution |
Refractive index | 1.4500 to 1.4540 (20°C, 589 nm) |
Assay | ≥50 % Solids |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...