2-Chloroethyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloroethyl carbonochloridate

39533-1000
TS39533-1000EA 165 USD
TS39533-1000
2-Chloroethyl chloroformate 97%
2-Chloroethyl chloroformate
Formula: C₃H₄Cl₂O₂
MW: 142.9687 g/mol
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –70 °C
Density: 1.39 g/cm³
Flash Pt: 70 °C
Storage Temperature: Refrigerator
UN: 3277
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR