(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 96+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

39223-0050 39223-0010
TS39223-0050EA 278.27 USD
TS39223-0050 TS39223-0010
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 96+%
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Formula: C₄₈H₄₀P₂
Melting Pt: 267…270 °C
Storage Temperature: Ambient
MDL Number: MFCD01311709
CAS Number: 153305-67-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR