Propargyl p-toluenesulfonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Propargyl p-toluenesulfonate
, Propargyl p-toluenesulphonate
37968-1000
37968-0250
37968-0050
TS37968-0050EA
46
USD
TS37968-0050
TS37968-0250
TS37968-1000
Propargyl p-toluenesulfonate 97%
Propargyl p-toluenesulfonate
Formula:
C₁₀H₁₀O₃S MW: 210.25 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00078365 CAS Number: 6165-76-0 |
Specification Test Results
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5270 to 1.5290 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...