2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonyl)imide
, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)
, Comins Triflating Reagent
, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine
, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide
, 2-[N,N-Bis(Trifluoromethylsulfonyl)amino]-5-chloropyridine
, 2-[N,N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine
, 2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine
, N-(5-Chloro-2-pyridyl)triflimide
Formula:
C₇H₃ClF₆N₂O₄S₂ MW: 392.67 g/mol Boiling Pt: 77…90 °C (.2 mmHg) Melting Pt: 45…47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191833 CAS Number: 145100-51-2 |
Specification Test Results
Appearance | White to light yellow Crystalline solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...