Dicyclohexylchlorophosphine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorodicyclohexylphosphine

35329-0050 35329-0010
TS35329-0010EA 70 USD
TS35329-0010 TS35329-0050
Dicyclohexylchlorophosphine 97%
Dicyclohexylchlorophosphine
Formula: C₁₂H₂₂ClP
Boiling Pt: 165 °C (12 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014294
CAS Number: 16523-54-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5310 to 1.5340 (20°C, 589 nm)
Titration Argentometric ≥96.0 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR