Cumene 99.9%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-PhenylpropaneIsopropylbenzene

32973-0025 32973-5000 32973-0050
TS32973-0050EA 34.51 USD
TS32973-0050 TS32973-5000 TS32973-0025
Cumene 99.9%, Extra Pure
Cumene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 152…154 °C (760 mmHg)
Melting Pt: –96 °C
Density: 0.862 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008881
CAS Number: 98-82-8
EINECS: 202-704-5
UN: 1918
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4930 (20°C, 589 nm)
Density (20°C) 0.861 to 0.863 kg/l
GC ≥99.85 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR