1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 0.97

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride

37831-0020
TS37831-0020EA 241.54 USD
TS37831-0020
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 0.97
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride
Formula: C₂₇H₃₇ClN₂
MW: 425.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684545
CAS Number: 250285-32-6

Order Now

Specification Test Results

Appearance White to off-white to tan Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR