1,2-Bis(diphenylphosphino)benzene 0.98

Supplier: Thermo Scientific Chemicals

36375-0010
TS36375-0010EA 87.59 USD
TS36375-0010
1,2-Bis(diphenylphosphino)benzene 0.98
1,2-Bis(diphenylphosphino)benzene
Formula: C₃₀H₂₄P₂
Melting Pt: 184…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00014081
CAS Number: 13991-08-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 184 to 189 °C
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR