4-Fluorophenyl chloroformate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluorophenyl carbonochloridate

30832-0050 30832-0010
TS30832-0050EA 91.3 USD
TS30832-0050 TS30832-0010
4-Fluorophenyl chloroformate 98%
4-Fluorophenyl chloroformate
Formula: C₇H₄ClFO₂
MW: 174.5569 g/mol
Flash Pt: 76 °C
Storage Temperature: Refrigerator
UN: 3277
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4923 to 1.4943 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR