Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane) 0.98

Supplier: Thermo Scientific Chemicals
Warning

27816-0050 27816-0010
TS27816-0010EA 109.88 USD
TS27816-0010 TS27816-0050
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane) 0.98
Dibenzo-21-crown-7 (Dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane)

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 104 to 108 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR