Dibenzyl azodicarboxylate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibenzyl diazene-1,2-dicarboxylate
27027-0250
27027-0050
TS27027-0050EA
58
USD
TS27027-0050
TS27027-0250
Dibenzyl azodicarboxylate 94%
Dibenzyl azodicarboxylate
Formula:
C₁₆H₁₄N₂O₄ MW: 298.3 g/mol Melting Pt: 44…52 °C Flash Pt: >230 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016737 CAS Number: 2449-05-0 EINECS: 219-508-0 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Yellow to orange Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 44 to 52 °C |
Loss on drying | ≤0.75 % |
UV spectrophotometry | ≥92 % |
Heavy metals (as Pb) | ≤30 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...