Tetrabutylammonium dihydrogentrifluoride 50-55% (w/w) in 1,2-dichloroethane

Supplier: Thermo Scientific Chemicals
Danger

26878-0025
TS26878-0025EA 114.14 USD
TS26878-0025
Tetrabutylammonium dihydrogentrifluoride 50-55% (w/w) in 1,2-dichloroethane
Tetrabutylammonium dihydrogentrifluoride
Formula: C₁₆H₃₈F₃N
Flash Pt: 13 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145365
CAS Number: 99337-56-1
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear light yellow to brown Solution
Infrared spectrum Conforms
NMR Consistent with structure
Strength 50 to 55 % (NMR)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR