Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzo-15-crown-5

24399-0025
TS24399-0025EA 96.3 USD
TS24399-0025
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99%
Benzo-15-crown-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula: C₁₄H₂₀O₅
MW: 268.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005945
CAS Number: 14098-44-3

Order Now

Specification Test Results

Appearance White to light grey Powder or flakes
Infrared spectrum Conforms
Melting point 79 to 82 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR