N-Methyl-bis-trifluoroacetamide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
, MBTFA
21007-0050
21007-0250
TS21007-0250EA
476
USD
TS21007-0250
TS21007-0050
N-Methyl-bis-trifluoroacetamide 98%
N-Methylbistrifluoroacetamide
Formula:
C₅H₃F₆NO₂ MW: 223.07 g/mol Boiling Pt: 120…122 °C (760 mmHg) Density: 1.547 g/cm³ Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000412 CAS Number: 685-27-8 EINECS: 211-680-5 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3450 to 1.3470 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...