Borane-tert-butylamine complex 95%, powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Boron hydride-tert-butylamine
17730-1000
17730-0250
TS17730-0250EA
65
USD
TS17730-0250
TS17730-1000
Borane-tert-butylamine complex 95%, powder
Borane tert-butylamine complex
Formula:
C₄H₁₄BN Melting Pt: 96…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075635 CAS Number: 7337-45-3 EINECS: 230-851-5 UN: 2926 ADR: 4.1,II |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 96 to 101 °C |
Titration | ≥94 % (Active hydrogen) |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...