Methyl (diethylphosphono)acetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-(diethoxyphosphoryl)acetate
, Diethyl methoxycarbonylmethanephosphonate
16998-0500
16998-0100
TS16998-0500EA
163
USD
TS16998-0500
TS16998-0100
Methyl (diethylphosphono)acetate 97%
Methyl (diethylphosphono)acetate
Formula:
C₇H₁₅O₅P MW: 210.17 g/mol Boiling Pt: 127…131 °C (9 mmHg) Density: 1.14 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009081 CAS Number: 1067-74-9 EINECS: 213-938-2 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4310 to 1.4350 (20°C, 589 nm) |
GC | ≥96.0 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...