Perchloroethane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexachloroethane

14854-1000 14854-0025 14854-0100 14854-0010
TS14854-0025EA 262 USD
TS14854-0025 TS14854-0100 TS14854-1000 TS14854-0010
Perchloroethane 99%
Perchloroethane
Formula: C₂Cl₆
MW: 236.74 g/mol
Melting Pt: 184…190 °C
Density: 2.091 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000799
CAS Number: 67-72-1
EINECS: 200-666-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 185 to 195 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR