1-(4-Nitrophenylazo)-2-naphthol, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Para Red1-((4-Nitrophenyl)diazenyl)naphthalen-2-ol

14630-0250
TS14630-0250EA 158 USD
TS14630-0250
1-(4-Nitrophenylazo)-2-naphthol, pure
1-(4-Nitrophenylazo)-2-naphthol
Formula: C₁₆H₁₁N₃O₃
MW: 293.28 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00003908
CAS Number: 6410-10-2
EINECS: 229-093-8

Order Now

Specification Test Results

Appearance Red Powder
Infrared spectrum Conforms
UV spectrum Complies
Elemental Analysis 29.03 to 31.45 % (C)
Elemental Analysis 11.29 to 12.23 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR