β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate 95+%, reduced

Supplier: Thermo Scientific Chemicals

Synonyms: beta-Nicotinamide adenine dinucleotide disodium salt hydratebeta-NADH disodium salt hydrateβ-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate

27110-2500 27110-0050 27110-0250 27110-0010
TS27110-0010EA 168 USD
TS27110-0010 TS27110-2500 TS27110-0050 TS27110-0250
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate 95+%, reduced
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
Formula: C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
MW: 727.43 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03791273
CAS Number: 606-68-8
EINECS: 210-123-3

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Water ≤8.0 % (K.F.)
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR