Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
, 4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate
16405-0010
16405-0050
TS16405-0050EA
450.9
USD
TS16405-0050
TS16405-0010
Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%
Bathophenanthrolinedisulphonic acid disodium salt hydrate
Formula:
C₂₄H₁₄N₂Na₂O₆S₂·H₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149310 CAS Number: 52746-49-3 EINECS: 258-152-0 |
Specification Test Results
Appearance | White to beige to pink Powder |
Infrared spectrum | Conforms |
Water | 2 to 6 % (K.F.) |
UV Lambda max | 276 to 284 nm (in water) |
Specific extinction E (1%/1cm) | ≥710 (at lambda max) |
Titration with HClO4 | 97.5 to 102.5 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...