1-Methylindole-3-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methylindole-3-boronic acid pinacol ester

44101-0050 44101-0010
TS44101-0050EA 832 USD
TS44101-0050 TS44101-0010
1-Methylindole-3-boronic acid pinacol ester 97%
1-Methylindole-3-boronic acid pinacol ester
Formula: C₁₅H₂₀BNO₂
Melting Pt: 106…112 °C
Storage Temperature: Ambient
MDL Number: MFCD11894338
CAS Number: 683229-61-0

Order Now

Specification Test Results

Appearance White to light orange to light brown Solid
Infrared spectrum Conforms
Melting point 105 to 113 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR