Methylmagnesium chloride ∼22% (w/w) 3 mol/l in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC25256-1000 25256-8000
TS25256-1000EA 65.12 USD
TS25256-1000 TS25256-8000
Methylmagnesium chloride ∼22% (w/w) 3 mol/l in THF, AcroSeal™
Methylmagnesium chloride
Formula: CH₃ClMg
Boiling Pt: 66 °C (760 mmHg)
Density: 1.01 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000468
CAS Number: 676-58-4
EINECS: 211-629-7
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Dark brown Solution
Titration with HCl 22 to 24 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR