Progesterone 98%

Supplier: Thermo Scientific Chemicals
Warning

22565-1000 22565-0050 22565-0250
TS22565-1000EA 320 USD
TS22565-1000 TS22565-0250 TS22565-0050
Progesterone 98%
Progesterone
Formula: C₂₁H₃₀O₂
Melting Pt: 126…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00003658
CAS Number: 57-83-0
EINECS: 200-350-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 131 °C
Loss on drying ≤0.5 % (500 mg, 105 °C, 2 hrs)
HPLC ≥97.5 %
Specific optical rotation +186° to +194° (20 °C, 589 nm) (c=1, ethanol) (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR