Isophorondiamine cis- and trans-mixture 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Amino-3-aminomethyl-3,5,5-trimethylcyclohexaneIsophoronediamine

35294-0010 35294-2500
TS35294-0010EA 116 USD
TS35294-0010 TS35294-2500
Isophorondiamine cis- and trans-mixture 99+%
Isophorondiamine
Formula: C₁₀H₂₂N₂
Boiling Pt: 247 °C
Melting Pt: 10 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00019397
CAS Number: 2855-13-2
EINECS: 220-666-8
UN: 2289
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4900 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤15 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR