Curcumin 98+% (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

21858-2500 21858-0500 21858-0100
TS21858-0500EA 236.21 USD
TS21858-0500 TS21858-0100 TS21858-2500
Curcumin 98+% (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)
Curcumin
Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
Melting Pt: 183 °C
Storage Temperature: Ambient
MDL Number: MFCD00008365
CAS Number: 458-37-7
EINECS: 207-280-5

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR