Triethylammonium chloride 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triethylamine hydrochloride

27067-0010 27067-2500
TS27067-2500EA 56.81 USD
TS27067-2500 TS27067-0010
Triethylammonium chloride 99+%
Triethylammonium chloride
Formula: C₆H₁₆ClN
MW: 137.65 g/mol
Melting Pt: 254…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00012500
CAS Number: 554-68-7
EINECS: 209-067-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 254 to 260 °C
Loss on drying ≤1.0 %
Solubility (10 % in water) Clear colorless
pH 4.5 to 6.5 (1 % w/v solution)
Titration with HClO4 99.0 to 101.0 % (On anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR