Sodium thiocyanate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium rhodanide

19676-5000 19676-0025 AC19676-2500
TS19676-5000EA 95 USD
TS19676-5000 TS19676-0025 TS19676-2500
Sodium thiocyanate 98%, Extra Pure
Sodium thiocyanate
Formula: NaSCN
MW: 81.07 g/mol
Melting Pt: 287 °C
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 208-754-4
Merck Index: 12,09463

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Argentometric ≥97.5 % (on dried substance)
Loss on drying ≤2.5 %
pH 5.5 to 7.5 (5 % at 20°C)
Iron (Fe) ≤2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR