Octadecylamine 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-OctadecylamineStearylamine1-AminooctadecaneOctadecan-1-amine

12932-5000 12932-0010 12932-0050
TS12932-0050EA 334 USD
TS12932-0050 TS12932-5000 TS12932-0010
Octadecylamine 90% tech.
Octadecylamine
Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269.51 g/mol
Boiling Pt: 232 °C (32 mmHg)
Melting Pt: 50…60 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00008159
CAS Number: 124-30-1
EINECS: 204-695-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White Pastilles
Infrared spectrum Conforms
Melting point 50 to 60 °C
Water ≤0.5 %
GC ≥88 %
Titration with HCl ≥98 % (prim.amine MW 269)
Titration with HCl ≥2 % (sec.+ tert.amine MW 521)
Iodine value ≤4 g I/100 g
Amine number 204 to 211 mg KOH/g (Total)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR