Aluminium acetate basic 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Aluminium Acetate Monobasic
AC42819-0010
AC42819-2500
TS42819-0010EA
168
USD
TS42819-0010
TS42819-2500
Aluminium acetate basic 90%
Aluminium acetate basic
Formula:
C₄H₇AlO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00008688 CAS Number: 142-03-0 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Aluminium (Al) | 15.0 to 18.3 % (ICP) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...