(3-Aminopropyl)dimethylmethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Danger

AC33660-0010 AC33660-0050
TS33660-0010EA 158.01 USD
TS33660-0010 TS33660-0050
(3-Aminopropyl)dimethylmethoxysilane 97%
(3-Aminopropyl)dimethylmethoxysilane
Formula: C₆H₁₇NOSi
Boiling Pt: 87 °C (50 mmHg)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD01861689
CAS Number: 31024-26-7
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR