4-Fluorphenylmagnesium bromide 0.8 mol/l in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluorophenylmagnesium bromide

AC37723-8000 AC37723-1000
TS37723-1000EA 54.41 USD
TS37723-1000 TS37723-8000
4-Fluorphenylmagnesium bromide 0.8 mol/l in THF, AcroSeal™
4-Fluorphenylmagnesium bromide
Formula: C₆H₄BrFMg
Melting Pt: 5 °C
Density: 1.01 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00009667
CAS Number: 352-13-6
UN: 3399
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Clear to hazy yellow to dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 14 to 17 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR