Potassium hydrogen diiodate 99.85%, Extra Pure for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium bi-iodate

AC21147-1000 AC21147-5000
TS21147-1000EA 152 USD
TS21147-1000 TS21147-5000
Potassium hydrogen diiodate 99.85%, Extra Pure for analysis
Potassium hydrogen diiodate
Formula: HI₂KO₆
MW: 389.91
Storage Temperature: Ambient
MDL Number: MFCD00011400
CAS Number: 13455-24-8
EINECS: 236-650-9
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White to almost white Powder or crystals
Titration Iodometric 99.80 to 100.20 %
Sulfate (SO4) ≤50 ppm
Cl, ClO3, Br, BrO3 (as Cl) ≤200 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤20 ppm
Lead (Pb) ≤5 ppm
Sodium (Na) ≤50 ppm
Total nitrogen (as N) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR