Sodium-4-hydroxymercuriobenzoate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Hydroxymercuri)benzoic acid sodium salt

AC41207-0100 AC41207-0010
TS41207-0100EA 120 USD
TS41207-0100 TS41207-0010
Sodium-4-hydroxymercuriobenzoate 98%
Sodium 4-hydroxymercuriobenzoate
Formula: C₇H₅HgNaO₃
Melting Pt: >300 °C
Storage Temperature: Ambient
CAS Number: 138-85-2
EINECS: 205-340-5
UN: 2026
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex Hg)
NMR ≤20 % (HCl salt)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR