Niobium(V) ethanolate 99.95% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Niobium(V) ethoxide

AC31688-0050 AC31688-0500
TS31688-0050EA 93.66 USD
TS31688-0050 TS31688-0500
Niobium(V) ethanolate 99.95% (trace metals basis)
Niobium(V) ethanolate
Formula: C₁₀H₂₅NbO₅
Boiling Pt: 140…142 °C (.1 mmHg)
Melting Pt: 6 °C
Density: 1.268 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00015122
CAS Number: 3236-82-6
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to orange-brown Liquid
Total trace metal impurities ≤500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR