Triethylborane 1 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC17698-8000 AC17698-1000
TS17698-1000EA 115.29 USD
TS17698-1000 TS17698-8000
Triethylborane 1 M in THF, AcroSeal™
Triethylborane
Formula: C₆H₁₅B
Density: 0.86 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00009022
CAS Number: 97-94-9
EINECS: 202-620-9
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to amber Solution
Strength 0.95 to 1.30 M
Water 4.2 to 6.0 %
HPLC ≥95 %
pH 4.0 to 6.0
Specific optical rotation 165° to 178° (20°C, 589 nm) (1 % in water DS)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR