Lithium-tert-butanolate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lithium tert-butoxide
AC30122-0250
AC30122-5000
AC30122-1000
TS30122-1000EA
393
USD
TS30122-1000
TS30122-0250
TS30122-5000
Lithium-tert-butanolate 99%
Lithium tert-butanolate
Formula:
C₄H₉LiO Density: 0.89 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00050479 CAS Number: 1907-33-1 EINECS: 217-611-5 UN: 2925 ADR: 4.1,II |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Titration with HCl | ≥98.5 % |
Impurity | ≤1 % (LiOH + Li2CO3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...