Sodium methanesulfinate 85%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methanesulfinic acid, sodium saltMethanesulphinic acid, sodium salt

AC39296-0500 AC39296-0010 AC39296-0100
TS39296-0010EA 28.63 USD
TS39296-0010 TS39296-0100 TS39296-0500
Sodium methanesulfinate 85%
Sodium methanesulphinate
Formula: CH₃NaO₂S
MW: 102.09 g/mol
Melting Pt: 222…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00040392
CAS Number: 20277-69-4

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR