Bismuth (III) trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bismuth(III) trifluoromethanesulfonate
, Bismuth triflate
AC38759-0050
AC38759-0250
TS38759-0050EA
77.42
USD
TS38759-0050
TS38759-0250
Bismuth (III) trifluoromethanesulfonate 98%
Bismuth (III) trifluoromethanesulfonate
Formula:
C₃BiF₉O₉S₃ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093669 CAS Number: 88189-03-1 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Titration Complexometric | 31.0 to 33.0 % Bi |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...