Tin(IV) acetate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

AC38005-0010 AC38005-0050
TS38005-0010EA 58.23 USD
TS38005-0010 TS38005-0050
Tin(IV) acetate 98%, Extra Pure
Tin (IV) acetate
Formula: C₈H₁₂O₈Sn
Melting Pt: 231…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00014976
CAS Number: 2800-96-6
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder
Melting point 231 to 237°C
Gravimetry 32.6 to 34.3 % Sn After ignition to oxide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR