3-Picolyl chloride hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Chloromethyl)pyridine hydrochloride3-(Chloromethyl)-pyridinium chloride

AC13119-0050 AC13119-0250
TS13119-0050EA 47.63 USD
TS13119-0050 TS13119-0250
3-Picolyl chloride hydrochloride 99%
3-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 137…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥99.4 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR